ADVH303C355D AChemBlock

3-chloro-1-methyl-1H-pyrazole

  • MDL number MFCD09879994
  • Cas Number : 63425-54-7
  • Molecular Weight : 116.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-chloro-1-methyl-1h-pyrazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q69919
Availability details for
Quantity
  • Availability
  • Add to Cart