ADVH303C357E AChemBlock

1-cyclopropyl-1H-pyrazol-4-amine

  • MDL number MFCD16810881
  • Molecular Weight : 123.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-cyclopropyl-1h-pyrazol-4-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q69952
Availability details for
Quantity
  • Availability
  • Add to Cart