ADVH303C3659 AChemBlock

BAY41-4109,Racemic

  • Cas Number : 298708-79-9
  • Molecular Weight : 395.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/bay41-4109-racemic.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q70217
Availability details for
Quantity
  • Availability
  • Add to Cart