ADVH49B81E29 AChemBlock

4-Octyl itaconate

  • Cas Number : 3133-16-2
  • Molecular Weight : 241.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/4-octyl-itaconate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86749
Availability details for
Quantity
  • Availability
  • Add to Cart