ADVH49B81E2C AChemBlock

7H-dibenzo[de,g]imidazo[2,1-a]isoquinoline-7,8(7aH)-dione

  • Cas Number : 23749-58-8
  • Molecular Weight : 286.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/7h-dibenzo-de-g-imidazo-2-1-a-isoquinoline-7-8-7ah-dione.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86661
Availability details for
Quantity
  • Availability
  • Add to Cart