ADVH49B81E2E AChemBlock

3-Bromo-5-ethyl-2-fluorophenylboronic acid pinacol ester

  • Cas Number : 2121514-74-5
  • Molecular Weight : 347.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-5-ethyl-2-fluorophenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70746
Availability details for
Quantity
  • Availability
  • Add to Cart