ADVH49B81E2F AChemBlock

4-Fluorobenzofuran-7-boronic acid pinacol ester

  • Cas Number : 2121511-48-4
  • Molecular Weight : 280.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-fluorobenzofuran-7-boronic-acid-pinacol-ester-294010.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70700
Availability details for
Quantity
  • Availability
  • Add to Cart