ADVH49B81E31 AChemBlock

2-(Trans-4-hydroxycyclohexyl)-1h-isoindole-1,3(2h)-dione

  • Cas Number : 99337-98-1
  • Molecular Weight : 245.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-trans-4-hydroxycyclohexyl-1h-isoindole-1-3-2h-dione.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86662
Availability details for
Quantity
  • Availability
  • Add to Cart