ADVH49B81E32 AChemBlock

7-Oxo-7H-benzo[de]benz[4,5]imidazo[2,1-a]isoquinoline-3-carbonitrile

  • Cas Number : 98842-91-2
  • Molecular Weight : 295.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/7-oxo-7h-benzo-de-benz-4-5-imidazo-2-1-a-isoquinoline-3-carbonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86743
Availability details for
Quantity
  • Availability
  • Add to Cart