ADVH49B81E33 AChemBlock

2-chloro-4-methoxypyridine-3-carbonitrile

  • Cas Number : 98645-43-3
  • Molecular Weight : 168.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-4-methoxypyridine-3-carbonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70879
Availability details for
Quantity
  • Availability
  • Add to Cart