ADVH49B81E39 AChemBlock

2-(1-benzothiophen-5-yl)ethan-1-ol

  • Cas Number : 96803-30-4
  • Molecular Weight : 178.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-1-benzothiophen-5-yl-ethan-1-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72528
Availability details for
Quantity
  • Availability
  • Add to Cart