ADVH49B81E3D AChemBlock

1,6-dimethyl-1,2,3,4-tetrahydroquinolin-4-one

  • Cas Number : 959699-02-6
  • Molecular Weight : 175.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-6-dimethyl-1-2-3-4-tetrahydroquinolin-4-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72551
Availability details for
Quantity
  • Availability
  • Add to Cart