ADVH49B81E3E AChemBlock

5,6-difluoro-1H-indole-3-carboxylic acid

  • Cas Number : 959238-71-2
  • Molecular Weight : 197.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-6-difluoro-1h-indole-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72558
Availability details for
Quantity
  • Availability
  • Add to Cart