ADVH49B81E41 AChemBlock

tert-butyl 2-(2-bromoethyl)morpholine-4-carboxylate

  • Cas Number : 958026-64-7
  • Molecular Weight : 294.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-2-bromoethyl-morpholine-4-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72570
Availability details for
Quantity
  • Availability
  • Add to Cart