ADVH49B81E43 AChemBlock

8-bromo-6-nitroH-imidazo[1,2-a]pyridine hydrochloride

  • Cas Number : 957120-43-3
  • Molecular Weight : 262.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/8-bromo-6-nitroh-imidazo-1-2-a-pyridine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70884
Availability details for
Quantity
  • Availability
  • Add to Cart