ADVH49B81E46 AChemBlock

6-chloroH-imidazo[1,2-a]pyridine hydrochloride

  • Cas Number : 957035-24-4
  • Molecular Weight : 193.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-chloroh-imidazo-1-2-a-pyridine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70886
Availability details for
Quantity
  • Availability
  • Add to Cart