ADVH49B81E4C AChemBlock

(1R)-1-[5-(2,2,2-trifluoroethoxy)-2-pyridyl]ethanamine

  • Cas Number : 953780-33-1
  • Molecular Weight : 220.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/1r-1-5-2-2-2-trifluoroethoxy-2-pyridyl-ethanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86587
Availability details for
Quantity
  • Availability
  • Add to Cart