ADVH49B81E53 AChemBlock

3-bromo-2-(4-chlorophenyl)imidazo[1,2-a]pyrimidine

  • Cas Number : 947613-07-2
  • Molecular Weight : 308.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-2-4-chlorophenyl-imidazo-1-2-a-pyrimidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72690
Availability details for
Quantity
  • Availability
  • Add to Cart