ADVH49B81E59 AChemBlock

5-bromo-4-fluoropyridin-2-amine

  • Cas Number : 944401-69-8
  • Molecular Weight : 191

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-4-fluoropyridin-2-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72747
Availability details for
Quantity
  • Availability
  • Add to Cart