ADVH49B81E5C AChemBlock

7-bromo-1H-pyrido[3,4-b][1,4]oxazin-2-one

  • Cas Number : 943995-72-0
  • Molecular Weight : 229.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-bromo-1h-pyrido-3-4-b-1-4-oxazin-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72754
Availability details for
Quantity
  • Availability
  • Add to Cart