ADVH49B81E5E AChemBlock

Acebilustat

  • Cas Number : 943764-99-6
  • Molecular Weight : 481.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/acebilustat.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72354
Availability details for
Quantity
  • Availability
  • Add to Cart