ADVH49B81E65 AChemBlock

(3-benzyloxetan-3-yl)methanol

  • Cas Number : 936501-51-8
  • Molecular Weight : 178.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-benzyloxetan-3-yl-methanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72839
Availability details for
Quantity
  • Availability
  • Add to Cart