ADVH49B81E67 AChemBlock

2,4-Dimethoxypyrimidine-5-boronic acid pinacol ester

  • Cas Number : 936250-17-8
  • Molecular Weight : 284.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-4-dimethoxypyrimidine-5-boronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70819
Availability details for
Quantity
  • Availability
  • Add to Cart