ADVH49B81E69 AChemBlock

MK-4101

  • Cas Number : 935273-79-3
  • Molecular Weight : 493.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/mk-4101.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72355
Availability details for
Quantity
  • Availability
  • Add to Cart