ADVH49B81E6C AChemBlock

2-methylimidazo[1,2-a]pyridine

  • Cas Number : 934-37-2
  • Molecular Weight : 132.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-methylimidazo-1-2-a-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72858
Availability details for
Quantity
  • Availability
  • Add to Cart