ADVH49B81E6E AChemBlock

cis-4-fluorocyclohexan-1-amine hydrochloride

  • Cas Number : 932706-30-4
  • Molecular Weight : 153.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/cis-4-fluorocyclohexan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72909
Availability details for
Quantity
  • Availability
  • Add to Cart