ADVH49B81E73 AChemBlock

2-chloro-7,8-dihydro-1,7-naphthyridin-8-one

  • Cas Number : 930303-55-2
  • Molecular Weight : 180.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-7-8-dihydro-1-7-naphthyridin-8-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72926
Availability details for
Quantity
  • Availability
  • Add to Cart