ADVH49B81E75 AChemBlock

(1-isobutyl-1H-pyrazol-4-yl)boronic acid

  • Cas Number : 929094-25-7
  • Molecular Weight : 168

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-isobutyl-1h-pyrazol-4-yl-boronic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72946
Availability details for
Quantity
  • Availability
  • Add to Cart