ADVH49B81E79 AChemBlock

3-(pyridin-2-yl)-1H-pyrazol-5-amine

  • Cas Number : 92352-29-9
  • Molecular Weight : 160.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-pyridin-2-yl-1h-pyrazol-5-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86537
Availability details for
Quantity
  • Availability
  • Add to Cart