ADVH49B81E7B AChemBlock

N-(4-chlorophenyl)-1-Piperazinecarboxamide

  • Cas Number : 923242-63-1
  • Molecular Weight : 239.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-4-chlorophenyl-1-piperazinecarboxamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86708
Availability details for
Quantity
  • Availability
  • Add to Cart