ADVH49B81E7C AChemBlock

benzyl 4-bromophenylcarbamate

  • Cas Number : 92159-87-0
  • Molecular Weight : 306.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/benzyl-4-bromophenylcarbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70923
Availability details for
Quantity
  • Availability
  • Add to Cart