ADVH49B81E7D AChemBlock

1H,2H-pyrido[2,3-d]pyrimidin-2-one

  • Cas Number : 91996-77-9
  • Molecular Weight : 147.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1h-2h-pyrido-2-3-d-pyrimidin-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73028
Availability details for
Quantity
  • Availability
  • Add to Cart