ADVH49B81E81 AChemBlock

1-(trifluoromethyl)-3-isocyanato-5-methoxybenzene

  • Cas Number : 918525-00-5
  • Molecular Weight : 217.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-trifluoromethyl-3-isocyanato-5-methoxybenzene.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70925
Availability details for
Quantity
  • Availability
  • Add to Cart