ADVH49B81E83 AChemBlock

[5-(prop-1-yn-1-yl)pyridin-3-yl]boronic acid

  • Cas Number : 917471-30-8
  • Molecular Weight : 160.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-prop-1-yn-1-yl-pyridin-3-yl-boronic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73051
Availability details for
Quantity
  • Availability
  • Add to Cart