ADVH49B81E84 AChemBlock

ethyl 4-oxopyrrolidine-3-carboxylate hydrochloride

  • Cas Number : 916814-29-4
  • Molecular Weight : 193.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-4-oxopyrrolidine-3-carboxylate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73055
Availability details for
Quantity
  • Availability
  • Add to Cart