ADVH49B81E85 AChemBlock

2-bromo-4-fluoro-5-methylbenzonitrile

  • Cas Number : 916792-07-9
  • Molecular Weight : 214.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-bromo-4-fluoro-5-methylbenzonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70927
Availability details for
Quantity
  • Availability
  • Add to Cart