ADVH49B81E8A AChemBlock

1H-indazole-5-carboxylic acid hydrochloride

  • Cas Number : 915139-44-5
  • Molecular Weight : 198.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1h-indazole-5-carboxylic-acid-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70932
Availability details for
Quantity
  • Availability
  • Add to Cart