ADVH49B81E8E AChemBlock

methyl 5-bromo-2-oxo-1,2-dihydropyridine-4-carboxylate

  • Cas Number : 913836-17-6
  • Molecular Weight : 232.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-bromo-2-oxo-1-2-dihydropyridine-4-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73097
Availability details for
Quantity
  • Availability
  • Add to Cart