ADVH49B81E91 AChemBlock

(1-methoxycyclopropyl)methanol

  • Cas Number : 912346-81-7
  • Molecular Weight : 102.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-methoxycyclopropyl-methanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73119
Availability details for
Quantity
  • Availability
  • Add to Cart