ADVH49B81E94 AChemBlock

7-Chloro-5-[[(1S)-1-phenylethyl]thio]-thiazolo[4,5-d]pyrimidin-2-amine

  • Cas Number : 911820-08-1
  • Molecular Weight : 322.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/7-chloro-5-1s-1-phenylethyl-thio-thiazolo-4-5-d-pyrimidin-2-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86789
Availability details for
Quantity
  • Availability
  • Add to Cart