ADVH49B81E9B AChemBlock

5-chloro-7-methoxythieno[3,2-b]pyridine

  • Cas Number : 90690-91-8
  • Molecular Weight : 199.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-chloro-7-methoxythieno-3-2-b-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86628
Availability details for
Quantity
  • Availability
  • Add to Cart