ADVH49B81EA0 AChemBlock

methyl 1-(difluoromethyl)-2-oxo-pyridine-3-carboxylate

  • Cas Number : 899452-36-9
  • Molecular Weight : 203.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-1-difluoromethyl-2-oxo-pyridine-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73242
Availability details for
Quantity
  • Availability
  • Add to Cart