ADVH49B81EA3 AChemBlock

6,8-difluorotetralin-1-one

  • Cas Number : 895534-38-0
  • Molecular Weight : 182.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-8-difluorotetralin-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73273
Availability details for
Quantity
  • Availability
  • Add to Cart