ADVH49B81EAA AChemBlock

4-chloropyridine-3-carbonitrile

  • Cas Number : 89284-61-7
  • Molecular Weight : 138.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-chloropyridine-3-carbonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73301
Availability details for
Quantity
  • Availability
  • Add to Cart