ADVH49B81EAF AChemBlock

1-methyl-1H-imidazol-4-amine hydrochloride

  • Cas Number : 89088-69-7
  • Molecular Weight : 133.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-methyl-1h-imidazol-4-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73319
Availability details for
Quantity
  • Availability
  • Add to Cart