ADVH49B81EB0 AChemBlock

6-((6-Chloropyrimidin-4-yl)oxy)-N-(3-(trifluoromethyl)phenyl)-1-naphthamide

  • Cas Number : 890128-71-9
  • Molecular Weight : 443.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-6-chloropyrimidin-4-yl-oxy-n-3-trifluoromethyl-phenyl-1-naphthamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86682
Availability details for
Quantity
  • Availability
  • Add to Cart