ADVH49B81EB5 AChemBlock

4,6-Difluoro-3-hydroxy-1H-indazole

  • Cas Number : 887567-77-3
  • Molecular Weight : 170.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-6-difluoro-3-hydroxy-1h-indazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72426
Availability details for
Quantity
  • Availability
  • Add to Cart