ADVH49B81EB7 AChemBlock

5-chloro-3-methylpyridine-2-carboxylic acid

  • Cas Number : 886365-46-4
  • Molecular Weight : 171.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-chloro-3-methylpyridine-2-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70973
Availability details for
Quantity
  • Availability
  • Add to Cart