ADVH49B81EBC AChemBlock

2,3-dibromo-6-chloropyridine

  • Cas Number : 885952-16-9
  • Molecular Weight : 271.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-3-dibromo-6-chloropyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86570
Availability details for
Quantity
  • Availability
  • Add to Cart