ADVH49B81EBE AChemBlock

3,6-Dibromo-1H-indazole

  • Cas Number : 885521-84-6
  • Molecular Weight : 275.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-6-dibromo-1h-indazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72410
Availability details for
Quantity
  • Availability
  • Add to Cart